3-Amino-5-fluor-1-methyl-1H-indazol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-1-methyl-1H-indazol-3-amine

PC5686-1G
APOSPC5686-1GEA 319 EUR
APOSPC5686-1G
3-Amino-5-fluor-1-methyl-1H-indazol 98%
3-Amino-5-fluoro-1-methyl-1H-indazole
Formel: C₈H₈FN₃
Molecular Weight: 165,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09054760
CAS-Nummer: 171809-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR