8-Iodo-6-(trifluoromethyl)imidazo[1,5-a]pyrimidine 95%

Lieferant: Apollo Scientific
Gefahr

PC56872-1G PC56872-250MG
APOSPC56872-1GEA 654 EUR
APOSPC56872-1G APOSPC56872-250MG
8-Iodo-6-(trifluoromethyl)imidazo[1,5-a]pyrimidine 95%
8-Iodo-6-(trifluoromethyl)imidazo[1,5-a]pyrimidine
Formel: C₇H₃F₃IN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31664911
CAS-Nummer: 2151834-40-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR