7-(Trifluoromethoxy)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC56889-1G PC56889-250MG
APOSPC56889-1GEA 864 EUR
APOSPC56889-1G APOSPC56889-250MG
7-(Trifluoromethoxy)-1H-indazole
7-(Trifluoromethoxy)-1H-indazole
Formel: C₈H₅F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD31664915
CAS-Nummer: 2244087-63-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR