6-(Difluoromethoxy)-1H-indazole ≥95%

Lieferant: Apollo Scientific
Achtung

PC56891-1G PC56891-250MG
APOSPC56891-1GEA 75.4 EUR
APOSPC56891-1G APOSPC56891-250MG
6-(Difluoromethoxy)-1H-indazole ≥95%
6-(Difluoromethoxy)-1H-indazole
Formel: C₈H₆F₂N₂O
Molecular Weight: 184.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22552662
CAS-Nummer: 1315359-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR