3-Bromo-4-chloro-2-fluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC56902-1G PC56902-250MG PC56902-5G PC56902-10G
APOSPC56902-1GEA 551 EUR
APOSPC56902-1G APOSPC56902-250MG APOSPC56902-5G APOSPC56902-10G
3-Bromo-4-chloro-2-fluorobenzaldehyde
3-Bromo-4-chloro-2-fluorobenzaldehyde
Formel: C₇H₃BrClFO
Molecular Weight: 237.454 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28733866
CAS-Nummer: 1806838-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR