Ethyl 6,7-difluoro-8-methoxy-4-oxo-1H-quinoline-3-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

PC56905-1G PC56905-250MG
APOSPC56905-1GEA 654 EUR
APOSPC56905-1G APOSPC56905-250MG
Ethyl 6,7-difluoro-8-methoxy-4-oxo-1H-quinoline-3-carboxylate 95%
Ethyl 6,7-difluoro-8-methoxy-4-oxo-1H-quinoline-3-carboxylate
Formel: C₁₃H₁₁F₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD31742295
CAS-Nummer: 950726-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR