3,4-Difluoro-5-nitrobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC56907-1G
APOSPC56907-1GEA 906 EUR
APOSPC56907-1G
3,4-Difluoro-5-nitrobenzoic acid 95%
3,4-Difluoro-5-nitrobenzoic acid
Formel: C₇H₃F₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD15144672
CAS-Nummer: 1121583-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR