3,4-Difluoro-5-nitrobenzonitrile 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyano-4,5-difluorobenzoic acid

PC56908-250MG
APOSPC56908-250MGEA 68.3 EUR
APOSPC56908-250MG
3,4-Difluoro-5-nitrobenzonitrile 95%
3,4-Difluoro-5-nitrobenzonitrile
Formel: C₈H₃F₂NO₂
Molecular Weight: 183,112 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11110377
CAS-Nummer: 1119454-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR