5-Bromo-2-cyano-3-fluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC56938-1G
APOSPC56938-1GEA 212 EUR
APOSPC56938-1G
5-Bromo-2-cyano-3-fluorobenzenesulphonyl chloride
5-Bromo-2-cyano-3-fluorobenzenesulphonyl chloride
Formel: C₇H₂BrClFNO₂S
Molecular Weight: 298,517 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28735185
CAS-Nummer: 1804387-41-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR