2-Bromo-4-chloro-6-fluorobenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC56942-1G PC56942-250MG PC56942-5G
APOSPC56942-1GEA 98.6 EUR
APOSPC56942-1G APOSPC56942-250MG APOSPC56942-5G
2-Bromo-4-chloro-6-fluorobenzaldehyde 95%
2-Bromo-4-chloro-6-fluorobenzaldehyde
Formel: C₇H₃BrClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD12400895
CAS-Nummer: 1135531-73-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR