2,6-Dimethyl-4-fluoranilin 98%

Lieferant: Apollo Scientific
Achtung

PC5695-1G PC5695-5G PC5695-25G PC5695-100G
APOSPC5695-1GEA 0 EUR
APOSPC5695-1G APOSPC5695-5G APOSPC5695-25G APOSPC5695-100G
2,6-Dimethyl-4-fluoranilin 98%
2,6-Dimethyl-4-fluoranilin
Formel: C₈H₁₀FN
Lagertemperatur: Kühlschrank
MDL: MFCD03792669
CAS-Nummer: 392-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR