2-Amino-3-(difluoromethyl)pyridine 98%

Lieferant: Apollo Scientific
Achtung

PC56951-1G PC56951-250MG PC56951-5G
APOSPC56951-1GEA 445 EUR
APOSPC56951-1G APOSPC56951-250MG APOSPC56951-5G
2-Amino-3-(difluoromethyl)pyridine 98%
2-Amino-3-(difluoromethyl)pyridine
Formel: C₆H₆F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19213737
CAS-Nummer: 878804-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR