2-Bromo-6-chloro-4-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC56967-1G PC56967-250MG PC56967-5G
APOSPC56967-1GEA 0 EUR
APOSPC56967-1G APOSPC56967-250MG APOSPC56967-5G
2-Bromo-6-chloro-4-(trifluoromethyl)benzoic acid 95%
2-Bromo-6-chloro-4-(trifluoromethyl)benzoic acid
Formel: C₈H₃BrClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28991853
CAS-Nummer: 1956322-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR