5-Chloro-4-fluoro-2-(trifluoromethyl)phenacyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-1-[5-chloro-4-fluoro-2-(trifluoromethyl)phenyl]ethanone
PC56994-1G
APOSPC56994-1GEA
654
EUR
APOSPC56994-1G
5-Chloro-4-fluoro-2-(trifluoromethyl)phenacyl bromide 95%
5-Chloro-4-fluoro-2-(trifluoromethyl)phenacyl bromide
Formel:
C₉H₄BrClF₄O Molecular Weight: 319,48 g/mol |
MDL:
MFCD28752704 CAS-Nummer: 1805956-11-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...