Methyl 3,5-difluoropyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluoro-2-(methoxycarbonyl)pyridine, Methyl 3,5-difluoropicolinate

PC57013-1G
APOSPC57013-1GEA 58 EUR
APOSPC57013-1G
Methyl 3,5-difluoropyridine-2-carboxylate
Methyl 3,5-difluoropyridine-2-carboxylate
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11616886
CAS-Nummer: 955885-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR