Methyl 3-bromo-5-fluoropyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-bromo-5-fluoropicolinate, 3-Bromo-5-fluoro-2-(methoxycarbonyl)pyridine

PC57016-250MG
APOSPC57016-250MGEA 159 EUR
APOSPC57016-250MG
Methyl 3-bromo-5-fluoropyridine-2-carboxylate
Methyl 3-bromo-5-fluoropyridine-2-carboxylate
Formel: C₇H₅BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14698117
CAS-Nummer: 1214337-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR