Methyl 6-chloro-5-fluoronicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-fluoro-5-(methoxycarbonyl)pyridine, Methyl 6-chloro-5-fluoropyridine-3-carboxylate

PC57018-1G
APOSPC57018-1GEA 539 EUR
APOSPC57018-1G
Methyl 6-chloro-5-fluoronicotinate
Methyl 6-chloro-5-fluoronicotinate
Formel: C₇H₅ClFNO₂
Molecular Weight: 189.572 g/mol
Schmelzpunkt: 87…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14698136
CAS-Nummer: 78686-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR