Methyl 5-chloro-3-fluoropyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-chloro-3-fluoropicolinate, 5-Chloro-3-fluoro-2-(methoxycarbonyl)pyridine

PC57019-250MG
APOSPC57019-250MGEA 26.1 EUR
APOSPC57019-250MG
Methyl 5-chloro-3-fluoropyridine-2-carboxylate
Methyl 5-chloro-3-fluoropyridine-2-carboxylate
Formel: C₇H₅ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14698145
CAS-Nummer: 1200498-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR