2-(Methylsulphonyl)-5-(trifluoromethyl)benzonitrile

Lieferant: Apollo Scientific
Achtung

PC57024-1G PC57024-5G PC57024-10G
APOSPC57024-1GEA 0 EUR
APOSPC57024-1G APOSPC57024-5G APOSPC57024-10G
2-(Methylsulphonyl)-5-(trifluoromethyl)benzonitrile
2-(Methylsulphonyl)-5-(trifluoromethyl)benzonitrile
Formel: C₉H₆F₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD21606312
CAS-Nummer: 1361385-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR