2',5'-Difluoro-2-(trifluoromethylsulphonyl)acetophenone
Lieferant: Apollo Scientific
Synonyme:
2-(2,5-Difluorophenyl)-2-oxoethyl trifluoromethyl sulphone, 1-(2,5-Difluorophenyl)-2-[(trifluoromethyl)sulphonyl]ethan-1-one, 2,5-Difluoro-3-{[(trifluoromethyl)sulphonyl]acetyl}benzene
PC57028-1G
APOSPC57028-1GEA
446
EUR
APOSPC57028-1G
2',5'-Difluoro-2-(trifluoromethylsulphonyl)acetophenone
2',5'-Difluoro-2-(trifluoromethylsulphonyl)acetophenone
Formel:
C₉H₅F₅O₃S Molecular Weight: 288,191 g/mol Schmelzpunkt: 57…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD20489421 CAS-Nummer: 1346521-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...