2-Bromo-6-nitro-4-(trifluoromethoxy)phenol

Lieferant: Apollo Scientific
Achtung

PC57035-5G
APOSPC57035-5GEA 862 EUR
APOSPC57035-5G
2-Bromo-6-nitro-4-(trifluoromethoxy)phenol
2-Bromo-6-nitro-4-(trifluoromethoxy)phenol
Formel: C₇H₃BrF₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD27665001
CAS-Nummer: 1820740-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR