2-Fluor-5-nitrobenzylalkohol 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-5-nitrophenyl)methanol

PC5704-10G PC5704-5G
APOSPC5704-10GEA 152 EUR
APOSPC5704-10G APOSPC5704-5G
2-Fluor-5-nitrobenzylalkohol 95+%
2-Fluor-5-nitrobenzylalkohol
Formel: C₇H₆FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02683502
CAS-Nummer: 63878-73-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR