3-[3-(Trifluoromethyl)phenyl]propanal
Lieferant: Apollo Scientific
|
Achtung
|
PC57040-1G
APOSPC57040-1GEA
79.9
EUR
APOSPC57040-1G
3-[3-(Trifluoromethyl)phenyl]propanal
3-[3-(Trifluoromethyl)phenyl]propanal
Formel:
C₁₀H₉F₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD07772918 CAS-Nummer: 21172-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...