4-(Difluoromethyl)aniline hydrochloride

Lieferant: Apollo Scientific
Achtung

PC57048-1G PC57048-250MG
APOSPC57048-1GEA 0 EUR
APOSPC57048-1G APOSPC57048-250MG
4-(Difluoromethyl)aniline hydrochloride
4-(Difluoromethyl)aniline hydrochloride
Formel: C₇H₇F₂N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28145075
CAS-Nummer: 1980063-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR