2-Fluor-5-(trifluormethoxy)phenol

Lieferant: Apollo Scientific
Gefahr

PC5707-1G PC5707-5G PC5707-10G
APOSPC5707-1GEA 46.4 EUR
APOSPC5707-1G APOSPC5707-5G APOSPC5707-10G
2-Fluor-5-(trifluormethoxy)phenol
2-Fluor-5-(trifluormethoxy)phenol
Formel: C₇H₄F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115893
CAS-Nummer: 886498-03-9
UN: 1760
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR