Ethyl (3-bromo-2-fluoro-6-formylphenoxy)acetate
Lieferant: Apollo Scientific
|
Achtung
|
PC57078-250MG
APOSPC57078-250MGEA
143
EUR
APOSPC57078-250MG
Ethyl (3-bromo-2-fluoro-6-formylphenoxy)acetate
Ethyl (3-bromo-2-fluoro-6-formylphenoxy)acetate
Formel:
C₁₁H₁₀BrFO₄ Molecular Weight: 305,097 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31922520 CAS-Nummer: 2366994-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...