5-Fluor-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
PC5711-1G
PC5711-250MG
APOSPC5711-1GEA
29
EUR
APOSPC5711-1G
APOSPC5711-250MG
5-Fluor-1H-indazol
5-Fluoro-1H-indazole
Formel:
C₇H₅FN₂ Schmelzpunkt: 119…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972877 CAS-Nummer: 348-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...