2-Bromo-4-chloro-5-fluorophenol ≥95%

Lieferant: Apollo Scientific

PC57127-250MG PC57127-1G
APOSPC57127-250MGEA 183 EUR
APOSPC57127-250MG APOSPC57127-1G
2-Bromo-4-chloro-5-fluorophenol ≥95%
2-Bromo-4-chloro-5-fluorophenol
Formel: C₆H₃BrClFO
MDL: MFCD26895908
CAS-Nummer: 157165-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR