5-Bromo-1-fluoro-2-iodo-3-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific
Achtung

PC57130-250MG PC57130-1G PC57130-5G
APOSPC57130-250MGEA 125 EUR
APOSPC57130-250MG APOSPC57130-1G APOSPC57130-5G
5-Bromo-1-fluoro-2-iodo-3-(trifluoromethyl)benzene 95%
5-Bromo-1-fluoro-2-iodo-3-(trifluoromethyl)benzene
Formel: C₇H₂BrF₄I
MDL: MFCD33404151
CAS-Nummer: 2385351-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR