2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester 98%

Lieferant: Apollo Scientific

PC5715-100MG PC5715-500MG
APOSPC5715-100MGEA 52.6 EUR
APOSPC5715-100MG APOSPC5715-500MG
2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester 98%
2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester
Formel: C₁₅H₁₉BF₃NO₃
Lagertemperatur: Kühlschrank
MDL: MFCD12026072
CAS-Nummer: 1150271-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR