4-Bromo-2-fluoro-6-(trifluoromethyl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC57296-250MG
PC57296-1G
PC57296-5G
APOSPC57296-250MGEA
125
EUR
APOSPC57296-250MG
APOSPC57296-1G
APOSPC57296-5G
4-Bromo-2-fluoro-6-(trifluoromethyl)benzoic acid 95%
4-Bromo-2-fluoro-6-(trifluoromethyl)benzoic acid
Formel:
C₈H₃BrF₄O₂ |
MDL:
MFCD33404165 CAS-Nummer: 2091679-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...