Sodium-3-fluorobenzoate 95%

Lieferant: Apollo Scientific
Achtung

PC5732-100G PC5732-25G PC5732-500G
APOSPC5732-100GEA 0 EUR
APOSPC5732-100G APOSPC5732-25G APOSPC5732-500G
Sodium-3-fluorobenzoate 95%
Sodium-3-fluorobenzoate
Formel: C₇H₄FO₂Na
Lagertemperatur: Raumtemperatur
MDL: MFCD03788550
CAS-Nummer: 499-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR