1-Bromo-1,1-dichloro-2,2,2-trifluoroethane 96%
Lieferant: Apollo Scientific
PC5733-1G
APOSPC5733-1GEA
132
EUR
APOSPC5733-1G
1-Bromo-1,1-dichloro-2,2,2-trifluoroethane 96%
1-Bromo-1,1-dichloro-2,2,2-trifluoroethane
Formel:
C₂BrCl₂F₃ Molecular Weight: 231,827 g/mol Siedepunkt: 69…69,2 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01746275 CAS-Nummer: 354-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...