3H-Perfluoro(2-methylbut-2-ene) 97%
Lieferant: Apollo Scientific
PC5739-5G
APOSPC5739-5GEA
452
EUR
APOSPC5739-5G
3H-Perfluoro(2-methylbut-2-ene) 97%
3H-Perfluoro(2-methylbut-2-ene)
Formel:
C₅HF₉ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10565607 CAS-Nummer: 22692-37-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...