2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%

Lieferant: Apollo Scientific

PC5741-250MG
APOSPC5741-250MGEA 43.5 EUR
APOSPC5741-250MG
2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%
2-Phenyl-4-(trifluoromethyl)-1H-imidazole
Formel: C₁₀H₇F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10000861
CAS-Nummer: 33469-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR