1-Bromo-4-ethenyl-3-fluoro-2-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific
Gefahr

PC57422-1G
APOSPC57422-1GEA 654 EUR
APOSPC57422-1G
1-Bromo-4-ethenyl-3-fluoro-2-(trifluoromethyl)benzene 95%
1-Bromo-4-ethenyl-3-fluoro-2-(trifluoromethyl)benzene
Formel: C₉H₅BrF₄
Molecular Weight: 269,03 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD34181536
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR