4-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-[4-bromo-2-fluoro-3-(trifluoromethyl)phenyl]ethanone

PC57424-1G
APOSPC57424-1GEA 654 EUR
APOSPC57424-1G
4-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide 95%
4-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide
Formel: C₉H₄Br₂F₄O
Molecular Weight: 363.93 g/mol
MDL: MFCD34181535
CAS-Nummer: 2386358-17-2
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR