4-Bromo-2-methoxy-3-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC57436-250MG PC57436-1G PC57436-5G
APOSPC57436-250MGEA 174 EUR
APOSPC57436-250MG APOSPC57436-1G APOSPC57436-5G
4-Bromo-2-methoxy-3-(trifluoromethyl)benzoic acid 95%
4-Bromo-2-methoxy-3-(trifluoromethyl)benzoic acid
Formel: C₉H₆BrF₃O₃
CAS-Nummer: 2383654-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR