Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

PC57446-1G
APOSPC57446-1GEA 928 EUR
APOSPC57446-1G
Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate 95%
Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate
Formel: C₁₀H₆BrFO₂S
Molecular Weight: 289,12 g/mol
MDL: MFCD13180865
CAS-Nummer: 360576-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR