Methyl 6-bromo-4-fluorobenzo[b]thiophene-2-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

PC57448-1G
APOSPC57448-1GEA 928 EUR
APOSPC57448-1G
Methyl 6-bromo-4-fluorobenzo[b]thiophene-2-carboxylate 95%
Methyl 6-bromo-4-fluorobenzo[b]thiophene-2-carboxylate
Formel: C₁₀H₆BrFO₂S
Molecular Weight: 289,121 g/mol
MDL: MFCD11044572
CAS-Nummer: 154650-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR