4-Bromo-3-(difluoromethyl)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

PC57452-1G
APOSPC57452-1GEA 609 EUR
APOSPC57452-1G
4-Bromo-3-(difluoromethyl)benzyl alcohol 95%
4-Bromo-3-(difluoromethyl)benzyl alcohol
Formel: C₈H₇BrF₂O
CAS-Nummer: 886860-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR