Methyl 2,3-difluoro-6-(trifluoromethoxy)benzoate 95%

Lieferant: Apollo Scientific
Achtung

PC57464-1G
APOSPC57464-1GEA 574 EUR
APOSPC57464-1G
Methyl 2,3-difluoro-6-(trifluoromethoxy)benzoate 95%
Methyl 2,3-difluoro-6-(trifluoromethoxy)benzoate
Formel: C₉H₅F₅O₃
Molecular Weight: 256,13 g/mol
CAS-Nummer: 2703797-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR