2-Bromo-1-chloro-5-fluoro-3-iodobenzene 95%

Lieferant: Apollo Scientific
Achtung

PC57468-1G
APOSPC57468-1GEA 654 EUR
APOSPC57468-1G
2-Bromo-1-chloro-5-fluoro-3-iodobenzene 95%
2-Bromo-1-chloro-5-fluoro-3-iodobenzene
Formel: C₆H₂BrClFI
Molecular Weight: 335,34 g/mol
CAS-Nummer: 2385641-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR