4-Chloro-5-fluoropyridine-3-carboxylic Acid 95%
Lieferant: Apollo Scientific
Achtung
|
PC57470-1G
APOSPC57470-1GEA
522
EUR
APOSPC57470-1G
4-Chloro-5-fluoropyridine-3-carboxylic Acid 95%
4-Chloro-5-fluoropyridine-3-carboxylic Acid
Formel:
C₆H₃ClFNO₂ Molecular Weight: 175,545 g/mol |
MDL:
MFCD18257385 CAS-Nummer: 860296-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...