3-(1,1-Difluoroethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC57526-250MG PC57526-1G PC57526-5G
APOSPC57526-250MGEA 190 EUR
APOSPC57526-250MG APOSPC57526-1G APOSPC57526-5G
3-(1,1-Difluoroethyl)benzoic acid 95%
3-(1,1-Difluoroethyl)benzoic acid
Formel: C₉H₈F₂O₂
Molecular Weight: 186.16 g/mol
MDL: MFCD20640021
CAS-Nummer: 55805-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR