4-Brom-2-fluor-5-methoxyphenylboronsäurepinakolester 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC5753-250MG
APOSPC5753-250MGEA
350
EUR
APOSPC5753-250MG
4-Brom-2-fluor-5-methoxyphenylboronsäurepinakolester 96%
4-Brom-2-fluor-5-methoxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BBrFO₃ Molecular Weight: 330,99 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11855986 CAS-Nummer: 1150271-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...