2-(3-Brom-4-(trifluormethoxy)phenyl)-2,2-difluoressigsäure 95%
Lieferant: Apollo Scientific
PC5754-1G
APOSPC5754-1GEA
510
EUR
APOSPC5754-1G
2-(3-Brom-4-(trifluormethoxy)phenyl)-2,2-difluoressigsäure 95%
2-(3-Brom-4-(trifluormethoxy)phenyl)-2,2-difluoressigsäure
Formel:
C₉H₄BrF₅O₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD11855929 CAS-Nummer: 1133116-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...