3H,3H-Perfluoro(2-methylbut-1-ene)

Lieferant: Apollo Scientific

PC5757-5G
APOSPC5757-5GEA 673 EUR
APOSPC5757-5G
3H,3H-Perfluoro(2-methylbut-1-ene)
3H,3H-Perfluoro(2-methylbut-1-ene)
Formel: C₅H₂F₈
Lagertemperatur: Raumtemperatur
MDL: MFCD12026455
CAS-Nummer: 400-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR