Diphenyl(4-methylphenyl)sulphonium triflate

Lieferant: Apollo Scientific
Gefahr

PC5762-1G
APOSPC5762-1GEA 0 EUR
APOSPC5762-1G
Diphenyl(4-methylphenyl)sulphonium triflate
Diphenyl(4-methylphenyl)sulphonium triflate

Photoinitiator/Photoacid Generator

Formel: C₁₉H₁₇S·CF₃O₃S
Molecular Weight: 426.472 g/mol
Schmelzpunkt: 101…102 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02683564
CAS-Nummer: 81416-37-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR