1-Ethyl-3-methylimidazoliumtrifluormethansulfonat

Lieferant: Apollo Scientific

Synonyme: 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

PC5766-25G
APOSPC5766-25GEA 89.9 EUR
APOSPC5766-25G
1-Ethyl-3-methylimidazoliumtrifluormethansulfonat
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate
Formel: C₇H₁₁F₃N₂O₃S
Molecular Weight: 260.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210008
CAS-Nummer: 145022-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR